Bel-Art™ SP Scienceware™ Pipettor Accuracy Test Kit
For most air displacement pipettors including Gilson, Rainin, Eppendorf, Oxform, SMI, Clinicon, Brand, MLA, Finn and other types
Manufacturer: Bel-Art™ 378700010
 View more versions of this product
Catalog No. 22-261793
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Pipettor accuracy test kit,Pipettor accuracy test kit
    Accuracy 0.005cm
    For Use With (Equipment) For air displacement pipettors
    For Use With (Application) For pipettor accuracy
    Volume (Metric) 10μL
    Includes Two flexible Vinyl adapters, 25 sets of Glass capillaries, 2 Glass pipettes for larger volumes
    Quantity 250 Pack
  • Description & Specifications
Related Products
Product Content Feedback