Learn More
Bel-Art™ SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station
SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station offers instant access to fast, safe, and effective first aid for eye and skin injuries.
Manufacturer:  Bel-Art™ F248805002
 View more versions of this product
Catalog No. 10-029-235
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
16 oz.
16 oz., 500mL
For workplaces where there is a risk of eye injuries from Acids or Alkalis
Wall
5 min./bottle
5 to 35°C
500mL
ANSI Z358.1-2009, U.S. federal regulations
Two 500mL bottles of saline, instructional pictogram, mirror and form fitting eye cups
Covered double eyewash station
Yes
Safety and Handling
ShelfLife : 3 Years