Learn More
Bel-Art™ SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station
SP Scienceware™ PLUM™ Emergency Covered Eye Wash Station offers instant access to fast, safe, and effective first aid for eye and skin injuries.
Manufacturer:  Bel-Art™ F248805003X
 View more versions of this product
Catalog No. 10-029-237
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
16 oz. and 6.76 oz.
16, 6.76 oz., 500mL, 200mL
For workplaces where there is a risk of eye injuries from Acids or Alkalis
Wall
5 min./500mL Bottle and 2 min./200mL bottle
5 to 35°C
500mL and 200mL
ANSI Z358.1-2009, U.S. federal regulations
One 500mL bottle of salen and one 200mL bottle pH neutralizing solution, instructional pictogram, mirror and form fitting eye cups
Covered double eyewash station
Yes
Safety and Handling
ShelfLife : 3 Years