Bel-Art™ SP Scienceware™ PLUM™ Emergency Eyewash Station Refills

Refill kits for PLUM station

Manufacturer: Bel-Art™ F248801023

 View more versions of this product

Catalog No. 10-029-231

  Check Availability
Add to Cart

    Specifications

    Product Type pH neutralizing eyewash solution refill
    Capacity (English) 6.76 oz.
    Capacity (Metric) 200mL
    Capacity 6.76 oz., 200mL
    For Use With (Equipment) PLUM eyewash stations or alone
    Includes Three 200mL bottles of sterile pH neutralizaing solution form-fitting eye cup, a dust cap and clear instructions
    Shelf Life 3 years

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit