Learn More
Bel-Art™ SP Scienceware™ PLUM™ Emergency Open Eye Wash Stations
SP Scienceware™ PLUM™ Emergency Open Eye Wash Stations provide instant access to fast, safe, and effective first aid for eye and skin injuries.
Manufacturer:  Bel-Art™ F248804010
 View more versions of this product
Catalog No. 19-991-001
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
33.81 oz.
33.81 oz., 1000mL
For dust-free workplaces where on the spot rinsing is required and mobile work spaces or smaller work spaces
30.5cm
Wall
10 min./bottle
5 to 35°C
1000mL
ANSI Z358.1-2009, U.S. federal regulations
12 in.
One 1000mL bottle of sterile saline, instructional pictogram and form fitting eye cup
Open single eyewash station
Yes
Safety and Handling
ShelfLife : 3 Years