Learn More
Bel-Art™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Manufacturer:  Bel-Art™ H131780068
 View more versions of this product
Catalog No. 01-815-13H
Add to cart
Description
Description
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
152mm
6 in.
PE (Polyethylene)
8.5 in.
0.1016mm
216mm
8.5 x 6 in. (216 x 152mm)