Learn More
Bel-Art™ Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Manufacturer:  Bel-Art™ H131761218
 View more versions of this product
Catalog No. 22-260172
Add to cart
Description
Description
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
18 x 12 in. (457 x 305mm)
0.038mm
457mm
305mm
18 in.
12 in.
PE (Polyethylene)