Learn More
Bel-Art Polyethylene Utility Bags
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Manufacturer: Bel-Art H131781216
Description
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
- FDA approved material; Latex free
- Different sizes available to best match particular packaging requirements
- Not autoclavable
Specifications
16 x 12 in. (406 x 305mm) | |
406mm | |
0.1016mm | |
305mm |
16 in. | |
PE (Polyethylene) | |
12 in. |