Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP SCIENCEWARE BEL ART SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring Available on GSA/VA Contract for Federal Government customers only.

Adds turbulence relative to similar smooth size cylindrical bars. SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars without Pivot Ring are manufactured in a registered ISO 9001:2008 facility.

Manufacturer:  SP SCIENCEWARE BEL ART F371200003

 View more versions of this product

Catalog No. 22-363325


  Check Availability
Add to cart

Specifications

Description

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility
  • Autoclave safe

Description & Specifications

Specifications

Yes
FDA, USP Class VI, ISO 9001:2008
White
Stir Bar
0.5 in.
12.7mm
50
PTFE
Alnico Magnet
Polygon
3 in.
76.2mm
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit