Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Polypropylene Left-Side Sink
Laboratory sink, drain board, backsplash and legs are made entirely of polypropylene plastic for excellent chemical resistance and easy cleaning
Supplier: Bel-Art™ H490110000
Description
- Material is unaffected by acids, alkalis and boiling solutions under 121°C (250°F)
- 15cm (6 in.) high backsplash and shorter side and front lip help to contain spills and channel liquids to the sink
- Made from 12.7mm (1/2 in.) thick material for durability
- Can contain approximately 20 gallons
- Sink is available on left or right side of unit
- Standard 1.5 in. “S” trap with plug is included to connect to plumbing drain
- Custom sizes or features such as longer units with double or oversize sinks and larger drain boards available
Specifications
20 gal. | |
76 L | |
Polypropylene | |
Left Side Sink |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title