Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 22-25mm

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Manufacturer:  Bel-Art™ 187740160

 View more versions of this product

Catalog No. 14-793-234



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Rack, Test Tube
Blue
11.62 x 7.37 x 3.25 in. (295 x 187 x 83mm)
83mm
11.62 in.
Epoxy Coated Steel Wire
7.37 in.
Yes
25mm
3.25 in.
Tubes (22 to 25mm)
295mm
60
187mm
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit