Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Pyrex™ Glass Round Spinbar™ Magnetic Stir Bars Available on GSA/VA Contract for Federal Government customers only.

Useful for high temperature applications up to 274°C (525°F)

Manufacturer:  Bel-Art™ F371010112

 View more versions of this product

Catalog No. 22-067579


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
USP Type I, ISO 9001:2008
Clear
Stir Bar
0.37 in.
9.5mm
50
Borosilicate Glass
Magnet
Round
1.5 in.
38mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit