Learn More
Bel-Art™ SP Scienceware™ Radioimmunoassay Tube Racks
Organize over 100 samples with one rack
$195.50 - $214.00
Specifications
Material Polypropylene
View More Specs
Products 3
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
14-809-11
View Documents
Bel-Art™
F188710000
Tubes (13 to 16 mm)
Each for $201.00
Case of 6 EA for $1,046.00
Add to cart
14-809-12
View Documents
Bel-Art™
F188720000
Tubes (16 to 20 mm)
Each for $214.00
Case of 6 EA for $1,114.00
Add to cart
14-809-10
View Documents
Bel-Art™
F188700000
Tubes (10 to 13 mm)
Each for $195.50
Case of 6 EA for $1,022.00
Add to cart
Description
Description
Organize over 100 samples with one rack
  • Polypropylene racks provide a practical method of pipetting and storing a large number of samples
  • Use alternate rows and rotate tubes to easily keep track of the last tube filled
  • Suitable for clinical lab use to store samples
  • Steam autoclavable at 121°C (250°F)
Specifications
Specifications
Polypropylene