Learn More
Bel-Art™ SP Scienceware™ Replacement Washers for the Pipette Pump™ Pipettors
Use with SP Scienceware™ Pipette Pump™ pipettors. Bel-Art™ SP Scienceware™ Replacement Washers are molded silicone rubber replacement parts. 3/Pack.
$27.00 - $29.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
22-261973
|
Bel-Art™
H937890081 |
10mL pipette pump pipettor |
Pack of 3 for $27.00
|
|
|||||
22-261975
|
Bel-Art™
H937890091 |
20mL pipette pump pipettor |
Pack of 3 for $29.00
|
|
|||||
Description
- For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
- Replacement washers to fit various sizes/colors of Pipette Pumps in case of loss or damage
- Molded silicone rubber is resistant to acids and alkalis
- Disassemble parts for cleaning and before autoclaving at 121°C (250°F) for 20 minutes
Specifications
Not Autoclavable | |
Washers |