Learn More
Bel-Art™ SP Scienceware™ Reusable Desiccant Cartridges Available on GSA/VA Contract for Federal Government customers only.
Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.
Manufacturer:  Bel-Art™ F420450160
 View more versions of this product
Catalog No. 08-594-17A
Add to cart
Description
Description
  • Desiccant changes color from blue when dry to pink when moist Saturated gel can be regenerated in a 150°C (300°F) vented oven until blue color returns
  • Cartridge can be regenerated repeatedly without degradation
  • Desiccant beads contained in a durable, polyester felt bag with a perforated Mylar° cover Cartridges are individually sealed in moisture proof pouches ready for immediate use
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
11.4cm
Polyester
Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads, for 160mm dia. desiccators
0.23kg
4.5 in.
Scienceware™ Reusable Desiccant Cartridge
0.5 lb.