Learn More
Bel-Art™ SP Scienceware™ Reuseable/Disposable Desiccant Cartridges Available on GSA/VA Contract for Federal Government customers only.
Use anywhere dessiccation is needed, including analytical balance cases and precision laboratory instruments. SP Scienceware™ Reuseable/Disposable Desiccant Cartridges fit “Space Saver” desiccators, in addition to other plastic desiccators of similar size.
Manufacturer:  Bel-Art™ F420480065
 View more versions of this product
Catalog No. 08-594-14A
Add to cart
Description
Description
  • Inert, noncorrosive and nontoxic silica gel
  • Clear polystyrene dish allows instant visual inspection of silica gel, which changes color from blue to pale pink as it absorbs moisture
  • Highly porous membrane covers the dish, providing excellent moisture absorption
  • Desiccant can be easily regenerated by heating cartridges in a 50°C (122°F) oven overnight
  • For more rapid drying, cut the membrane, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150°C (300°F) oven for one hour
  • Individually foil sealed
  • Fits “Space Saver” dessicator, in addition to other plastic dessicators of similar size
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
6.5cm
Aluminum
Desiccant Cartridge
2.55 in.
Yes
Use analytical balance cases and precision laboratory instruments