Bel-Art™ SP Scienceware™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position

Manufacturer: Bel-Art™ F124320050

 View more versions of this product

Catalog No. 03-411-741

  Check Availability
Add to Cart

    Specifications

    Chemical Label Assorted
    Color Assorted
    Capacity (Metric) 1000mL
    Capacity (English) 32 oz.
    Material LDPE
    Quantity 4/Pk.
    Capacity 32 oz., 1000mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit