Bel-Art™ SP Scienceware™ Right-to-Know, Safety-Vented Wash Bottles with GHS Labeling

Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position

Manufacturer: Bel-Art™ F124160003

 View more versions of this product

Catalog No. 03-411-722

  Check Availability
Add to Cart

    Specifications

    Chemical Label Deionized Water
    Capacity (Metric) 500mL
    Capacity (English) 16 oz.
    Color Blue
    Material LDPE
    Quantity 4/Pk.
    Capacity 16 oz., 500mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit