Learn More
Bel-Art™ SP Scienceware™ Rimless Centrifuge Tubes
For use in centrifuges up to 50,000xG without refrigeration
$47.50 - $79.07
Specifications
Autoclavable Autoclavable
Material Polypropylene
Max. Force 50,000 x g
Type Rimless Centrifuge Tube
No. per Case 12
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
22-260705
View Documents
Bel-Art™
F198070000
1.521 oz. 45 mL
Pack of 12 for $47.50
Case of 12 PK for $834.59
Add to cart
22-260706
View Documents
Bel-Art™
F198080000
3.04 oz. 90 mL
Pack of 12 for $79.07
Case of 12 PK for $3,171.20
Add to cart
Description
Description
  • Polypropylene
  • Round-bottomed
  • Autoclavable at 121°C (250°F)
Specifications
Specifications
Autoclavable
50,000 x g
12
Polypropylene
Rimless Centrifuge Tube
Round