Learn More
Bel-Art™ SP Scienceware™ Riteflow™ Unmounted Flowmeters Available on GSA/VA Contract for Federal Government customers only.
For corrosive and noncorrosive liquids and gases
Manufacturer:  Bel-Art™ H404030235
 View more versions of this product
Catalog No. 11-163-72G
  Check Availability
Add to cart
Alert:
Gaseous hydrofluoric acid may damage the glass.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
150mm Guarded
65.5°C
Glass and Stainless Steel
7.5 in.
200psi
±2%
Glass: 3 to 209.5; Stainless: 19 to 506.3mL/min.
19cm