Learn More
Bel-Art™ SP Scienceware™ Riteflow™ Unmounted Flowmeters
For corrosive and noncorrosive liquids and gases
Manufacturer:  Bel-Art™ H404010075
 View more versions of this product
Catalog No. 11-163-70C
  Check Availability
Add to cart
Alert:
Gaseous hydrofluoric acid may damage the glass.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Specifications Description & Specifications
Specifications
150mm Model with Plain Ends
121°C
Glass and Stainless Steel
7.5 in.
200psi
±2%
Glass: 0.23 to 5.5; Stainless: 1.1 to 20.4mL/min.
19cm