Learn More
Bel-Art™ ProCulture Round Microcentrifuge Floating Bubble Racks Available on GSA/VA Contract for Federal Government customers only.
Optimized for use in microbiology and immunology labs
$21.45 - $32.85
Specifications
Autoclavable Autoclavable
Material PMP
Type Microtube Rack
For Use With (Application) Incubating microtubes in water baths or beakers
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Holds Positions Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Holds Positions Diameter (Metric) Outer Price Quantity & Availability  
14-792-14 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Bel-Art™
F188750400
400mL Beakers Tubes (1.5 mL) 8 66 mm
Each for $28.50
Case of 12 EA for $276.00
Add to cart
14-792-15 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Bel-Art™
F188751000
1000mL Beakers Tubes (1.5 mL) 20 95 mm
Each for $32.85
Case of 12 EA for $309.50
Add to cart
13-717-272
View Documents
Bel-Art™
F188751600
1000mL Beakers Tubes (0.5 mL) 20 95 mm
Each for $21.45
Case of 12 EA for $230.50
Add to cart
Description
Description
  • Made especially for use with beakers and waterbaths, these space-saving racks are ideal for small quantities of tubes and for use in cell culture laboratories. 
  • Perfect for incubating microtubes in a beaker of water
  • Buoyant TPX™ brand polymethylpentene with tiny bubbles throughout for additional buoyancy
  • Insert tubes from the top and easily remove them by pressing the rack down on the bench
  • Withstands boiling for 30 minutes at 100°C (212°F)
  • Steam autoclavable at 121°C (250°F)
  • Leg length: 19mm (3/4 in.); thickness: 6.4mm (1/4 in.)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Eight-place disc measures 2.625 in. dia. (6.6cm); 20-place disc measures 3.75 in. dia. (9.5cm). Discs are 0.25 in. thick (0.6cm). Legs are 0.75 in. long (1.9cm).
Specifications
Specifications
Autoclavable
PMP
Microtube Rack
Incubating microtubes in water baths or beakers