Learn More
Bel-Art™ SP Scienceware™ Roxy-M™ Repeating Pipetter
Provides precise, accurate and reliable repetitive pipetting of volumes from 1µL to 5mL
$344.00
Specifications
Autoclavable Autoclavable
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
12-948-118
View Documents
Bel-Art™
F373100000
Roxy M Repeating Pipetter Each for $344.00
Add to cart
Description
Description
  • Ambidextrous design is slim, lightweight (105g) and ergonomic for a comfortable grip and smooth dispensing
  • Select the desired volume setting by turning the adjustment wheel, fill the tip by sliding the fill lever, and repeatedly dispense the desired volume in intervals as fast as one per second
  • Robust construction and positive displacement operation result in accurate and precise dispensing of a wide range of liquids, including high viscosity or high vapor pressure liquids
  • Includes adapter for use with 25 and 50mL tips (available separately, Mfr. No. 37300-0050)
  • Manual operation—no batteries required
  • Maintenance-free; No internal wear-and-tear parts
  • Includes instructions in English, French and Spanish
  • Identified with a serial number, tested for operability and precision, and comes with a Certificate of Conformity
Order Info
Use with Scienceware Roxy M sterile or nonsterile tips to offer nearly 40 different volume settings in as many as 48 steps without refilling the syringe tip.
Specifications
Specifications
Autoclavable
Polypropylene