Learn More
Bel-Art™ SP Scienceware™ Sample Cutter Set
One-piece polycarbonate tool for handling samples of semi-solid foods or materials
Manufacturer:  Bel-Art™ F369660000
Catalog No. 01-816-17
Add to cart
Description
Description
  • The three tined fork has a sharp rocker knife on its back edge and a strong handle
  • Sold in pairs so that one cutter can be used as a fork to hold the material on a flat surface or cutting board while the knife edge of the other tool is used in a rocking and sawing motion to cut off a sample
  • Samples can be picked up using the fork tines
  • The multifunctional tool is 19cm (7.5 in.) long
  • Autoclavable at 121°C (250°F) and safe to put into a commercial dishwasher
Order Info
2 per bag
Specifications
Specifications
Yes
Rocker knife and 3 tined fork on an easy to grip handle
19cm
Sample Cutter Set
Designed for handling samples of semi-solid foods or materials
7.5 in.
Polycarbonate