Learn More
Bel-Art™ SP Scienceware™ Sample Vials with Captive Closure
Friction-fit, captive closures are easy to open and close with a flick of the thumb
Manufacturer:  Bel-Art™ F175700000
 View more versions of this product
Catalog No. 22-171140
Add to cart
Description
Description
  • Cylindrical, low-density polyethylene (LDPE) plastic vials
  • Excellent for storage of liquids as well as for storing and shipping dry materials
Specifications
Specifications
Captive
13.7 mm
Cylindrical
0.43 oz.
Polypropylene
Use in cleanrooms and have excellent chemical resistance
144 Case
Sample Vial
7 mm
0.13 mL