Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Secador™ Auto Desiccator Cabinets: Horizontal Models Available on GSA/VA Contract for Federal Government customers only.

A UV light blocking, dry environment with automatic electronic dehumidification

Manufacturer:  Bel-Art™ F420740220

 View more versions of this product

Catalog No. 08-647-673


Add to cart

Description

Description

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626

  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
  • Dimensions
    • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in. H
    • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in. H
    • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in. H

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Order Info

    Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001

    Specifications

    Specifications

    Clear
    41.4cm
    Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
    Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
    Chemicals, stains and fine cracks
    41° to 104°F
    230V
    8.9 in.
    22.5cm
    Horizontal
    No
    34.1cm
    16.3 in.
    Bottom-hinged, pull down oven style
    16.4 in.
    4.0
    Vented adjustable shelves
    5° to 40°C
    1.9 cu. ft.
    20.4 in.
    51.8cm
    Copolyester
    13.4 in.
    2
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit