Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Secador™ Auto Desiccator Cabinets: Vertical Models Available on GSA/VA Contract for Federal Government customers only.

A UV light blocking, dry environment with automatic electronic dehumidification

Manufacturer:  Bel-Art™ F420731228

 View more versions of this product

Catalog No. 08-647-856


Add to cart

Description

Description

A UV light blocking, dry environment with automatic electronic dehumidification

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626

  • 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
  • Dimensions
    • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in. H
    • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in. H
    • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in. H

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Order Info

    Replacement shelves are available.

    Specifications

    Specifications

    Chemicals, stains and fine cracks
    41° to 104°F
    230V
    13.4 in.
    Vertical, tinted
    Amber
    41.4cm
    Side-swinging
    Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
    41.4cm
    No
    41.7cm
    Perforated shelves
    5° to 40°C
    1.55 cu. ft.
    34.1cm
    Copolyester
    16.3 in.
    16.3 x 13.4 in. (41.4 x 34.1cm)
    Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
    16.3 in.
    3.0
    16.4 in.
    3
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit