Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Models Available on GSA/VA Contract for Federal Government customers only.

A UV light-blocking, dry environment

Manufacturer:  Bel-Art™ F420740000

 View more versions of this product

Catalog No. 08-647-58


Add to cart

Description

Description

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50%, making them outstanding choices for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Easy-to-view hygrometer is located in the door front
  • 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3L x 13.4W x 8.4 in.H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in.H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in.H

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100.

Specifications

Specifications

Clear
41.4cm
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Chemicals, stains and fine cracks
1.9 cu. ft.
20.4 in.
51.8cm
Copolyester
13.4 in.
2
16.3 in.
Bottom-hinged and gasketed
16.4 in.
4.0
Adjustable perforated shelves
8.9 in.
41.7cm
Horizontal
No
34.1cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit