Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Model 1.0 Available on GSA/VA Contract for Federal Government customers only.

Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets block 99% of UV light and are resistant to staining, crazing and chemical attack.

Manufacturer:  Bel-Art™ F420710006

 View more versions of this product

Catalog No. 08-647-51


Add to cart

Description

Description

  • Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Stack up to 3 units high to save space
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Chemicals, stains and fine cracks
0.75 cu. ft.
13.4 in.
34.1cm
Copolyester
16.3 in.
16.3 x 13.4 in. (41.4 x 34.1cm)
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
Carrying handles, one latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer
41.4cm
No
21.3cm
Fixed perforated shelf
8.9 in.
22.5cm
Vertical
Clear with Blue End Caps
41.4cm
Left-hinged and gasketed
4.5 in.
16.3 in.
1.0
8.4 in.
1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit