Learn More
SP Bel-Art™ Secador™ Standard Desiccator Cabinets
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
Supplier: SP Bel-Art™ F420741006
Description
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
- Can be used with desiccant cartridges, F42049-0100 or F42046-0000
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. F42049-0100, F42046-0000
Specifications
Not Autoclavable | |
Vertical | |
Left-hinged and gasketed | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
3 | |
Removable Perforated | |
13.4 in. | |
34.1 cm | |
16.4 in. | |
16.3 in. |
Clear with Blue End Caps | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant | |
4.0 | |
Chemicals, stains and fine cracks | |
1.9 cu. ft. | |
8.9 in. | |
22.5 cm | |
51.8 cm | |
41.4 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.