Learn More
Bel-Art™ SP Scienceware™ Slide Staining and Storage Apparatus
Conveniently stain and store gels
Manufacturer:  Bel-Art™ F441800000
 View more versions of this product
Catalog No. 22-261920
Add to cart
Description
Description
  • Versatile slide rack and vessel for radiography gel staining and staining protocols
  • PMPA plastic construction makes the unit virtually shatterproof, lightweight, with exceptional solvent and heat resistance for extended use in cytology and histology labs
  • The rack and dish can simply be cleaned and are highly resistant to standard stains used in staining protocols
Specifications
Specifications
Black
Slide Rack
90°C
9cm
Used in cytology and histology labs
25
3.6 in.
3 in.