Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ SMORC™ Small Orifice Cleaner
Uniquely designed for cleaning and unclogging small orfices; a pin vise securely holds a stainless steel cleaning wire in place
Supplier: Bel-Art™ F381250000
Description
- 2.2cm (7/8 in.) diameter ring magnet on the handle is useful for picking up wires and for preventing the unit from rolling on metal surfaces
- Length without wire is 10.2cm (4 in.)
- Includes H38126-0000 wire assortment; stores conveniently inside the hollow handle
- Wire assortment contains 30 stainless steel wires, 5.1cm (2 in.) long, packaged in a plastic vial
- 10 each of 3 sizes: 0.152, 0.254, 0.508mm (0.006, 0.010, 0.020 in.)
Specifications
Small Orifice Cleaner | |
4 in. | |
Clean and clear small opening such as sample and reagent probe, reusable syringes, tubing |
Probe, reusable syringe, tubing | |
10.2 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction