Bel-Art™ SP Scienceware™ SMORC™ Small Orifice Cleaner Available on GSA/VA Contract for Federal Government customers only.
Uniquely designed for cleaning and unclogging small orfices; a pin vise securely holds a stainless steel cleaning wire in place
Manufacturer: Bel-Art™ H381260000
 View more versions of this product
Catalog No. 13-717-12
  Check Availability
Add to Cart
    Specifications
    Length (English) 2 in.
    Product Type Small orifice cleaner replacement wire
    Length (Metric) 5.1cm
    Material Stainless Steel
    For Use With (Application) Clean and clear small opening such as sample and reagent probe, reusable syringes, tubing
    For Use With (Equipment) Small orifice cleaner
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback