Learn More
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bars
SCI_ED
1.18 x 0.31 in. polygon magnetic stirring bar spins freely inside a stationary capsule, excellent for pH adjustments of small volumes.
Supplier:  Bel-Art™ F371520000/EMD
Catalog No. S96719
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Large, flat capsule can be used in vessels without «wandering«
  • Capsule is made of polystyrene and the magnetic stirring bar is PTFE-encapsulated
  • Magnetic stirring bar enclosed in capsule will not impact and damage expensive probes immersed in solution being stirred
  • Eliminate destructive forces that shakers and loose stirring bars can have on delicate gels by gently mixing destaining solutions with the Spinbar™ Capsule Magnetic Stirring Bar
Specifications
Specifications
53.3 mm
5 in.
127mm
Polygon
2.1 in.
Product Suggestions
Product Suggestions