Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainers Products for Science Education

Prevents Spinbar Magnetic Stirring Bars from going down the sink drain.

$44.25

Specifications

For Use With (Application) Prevents stir bar from washing down drain
For Use With (Equipment) Stirring bars
Includes 4 strong magnets in each corner, long slits in the sides and wide skirts
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
S04610 Products for Science Education
View Documents
Bel-Art™
F377870000/EMD
Blue Each for $44.25
Case of 12 EA for $386.05
Add to cart
 
S04611 Products for Science Education
View Documents
Bel-Art™
F377870001/EMD
Green Each for $44.25
Case of 12 EA for $378.44
Add to cart
 
Description

Description

  • Flexible, chemically resistant sink strainer has four strong magnets in each corner to attract and hold Spinbar Magnetic Stirring Bars and prevent them from going down the drain
  • Long slits in the sides and skirt allow fluids to drain easily and rapidly
  • A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
  • Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
  • Color choices improve visibility of stirring bars

Specifications

Specifications

Prevents stir bar from washing down drain
Stirring bars
4 strong magnets in each corner, long slits in the sides and wide skirts
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit