Bel-Art™ SP Scienceware™ Stack Rack Test Tube Racks

Feature interlocking end plates with handles for secure stacking.

Manufacturer: Bel-Art™ F188601620

 View more versions of this product

Catalog No. 11-391-306

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Holds For 16 to 20mm tubes
    Positions 40 places
    Product Type Test Tube Racks
    Material Polypropylene
    Tube Size 16-20mm

    • Polypropylene rack
    • Sturdy and chemical resistant
    • For tubes up to 10cm (4 in.) long
    • Numbered and lettered rows for easy tube identification
    • Steam autoclavable at 121°C (250°F)
    • With white surface to make color comparisons easy
    • Does not shatter or stain

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit