Learn More
Bel-Art™ SP Scienceware™ Sterileware™ Bent Handle Scoops
SCI_ED
Features an ergonomic, curved handle design to elevate hand from the sample, reducing risk of contamination and minimizing wrist strain.
$39.00
Specifications
Bottom Shape Flat
Certifications/Compliance FDA
Product Type Sampling scoop
Disposable Yes
For Use With (Application) Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Price Quantity & Availability  
S04787
SCI_ED
View Documents
Bel-Art™
H369022006/EMD
2 oz., 60mL 7.5 x 1.62 x 1.5 in. (19.1 x 4.1 x 3.7cm)
Pack of 10 for $39.00
Case of 10 PK for $390.00
Add to cart
Description
Description
  • Made from high-impact, lightweight, FDA grade polystyrene
  • Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
  • Flat bottom keeps scoop upright when placed on flat surface, can be used as weigh boat
  • Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with Certificate of Sterilization
  • Individually packaged and ready for one time use and disposal
  • Pack of 10
Specifications
Specifications
Flat
Sampling scoop
Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
Certificate of processing for sterilization
19.1cm
Sterile
FDA
Yes
Bent and curved
7.5 in.
Polystyrene