Bel-Art™ SP Scienceware™ Sterileware™ Economy Sample Spoons Available on GSA/VA Contract for Federal Government customers only.

Molded high-impact polystyrene for durability and stiffness. SP Scienceware™ Sterileware™ Economy Sample Spoons are ideal for retrieving small, specific volume samples of powders or liquids.

Manufacturer: Bel-Art™ F367400004

 View more versions of this product

Catalog No. 03-410-634

  Check Availability
Add to Cart

    Specifications

    Disposable Yes
    Includes Certificate of processing for sterilization
    Sterile Yes
    Capacity (English) 0.27 oz.
    Capacity (Metric) 8mL
    Product Type Sample Spoon
    Bottom Shape Flat
    Capacity 0.27 oz., 8mL
    Certifications/Compliance FDA
    Color White
    For Use With (Application) Designed for retrieving small, specific volume samples of powders or liquids and also provides durability and stiffness when sampling packed powders
    Material Polystyrene

    • Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
    • Flat-bottom shape permits use as a weighing boat
    • For one-time use

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit