Learn More
Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
Manufacturer:  Bel-Art™ F377520000
 View more versions of this product
Catalog No. 14-511-20
Add to cart
Description
Description
  • Up and down motion in cell provides complete mixing
  • Shaft diameter 3.17mm (0.125 in.)
  • Straight handle
Used to add enzymes and other materials to solution in spectrophotometer cells.
Specifications
Specifications
Cell type: 10mm light path; Straight handle
Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
36/Cs.
11.4cm
0.125 in.
4.5 in.
3.17mm
Acrylic