Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer

Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents

Manufacturer:  Bel-Art™ F377520001

 View more versions of this product

Catalog No. 14-511-21


Add to cart

Description

Description

  • Up and down motion in cell provides complete mixing
  • Shaft diameter 3.17mm (0.125 in.)
  • Straight handle

Used to add enzymes and other materials to solution in spectrophotometer cells.

Specifications

Specifications

36/Cs.
13cm
Cell type: Semi-micro; Straight handle
Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
3.17mm
Acrylic
0.125 in.
5.1 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit