Bel-Art™ SP Scienceware™ Swing Jaw™ Tubing Clamps
Provide nonslip closures for flexible tubing
Manufacturer: Bel-Art™ H182200000
 View more versions of this product
Catalog No. 05-833
  Check Availability
Add to Cart
    Specifications
    Product Type Tubing Clamp
    For Use With (Equipment) 10mm (3/8 in.) O.D. flexible tubing
    Jaw Type Swing
    Material Zinc-plated steel
  • Description & Specifications
  • Accessories
  • Promotions
Related Products
Product Content Feedback