Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Table-Top Buchner Funnels

For filtering large volumes quickly.

Manufacturer:  Bel-Art™ H146231914

 View more versions of this product

Catalog No. 14-100-128


  Check Availability
Add to cart

Specifications

Description

  • Polyethylene plastic
  • Removable hose barbed connector accepts 0.5 in. I.D. tube
  • Threads into 0.75 in. NPT bung on funnel
Description & Specifications

Specifications

36 in.
14.75 in.
Polyethylene
Coarse
91.4cm
37.5cm
with Coarse Porosity Removable Plate
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit