Learn More
Bel-Art™ SP Scienceware™ Techni-Dome™ 360 Gas-Ported Desiccator
For contamination and dust-free desiccation, storage and degassing
Manufacturer:  Bel-Art™ F420290001
Catalog No. 14-513-202
Add to cart
Description
Description
  • Protects generous quantities of standards, samples, electronics or other moisture-sensitive items
  • Spacious 65 liter (4000 cu. in.) internal volume can hold items up to 45cm (18 in.) tall or wide, including racks, trays and instruments
  • Two gas ports are tapered and accept tubing with 1/4 to 3/8 in. I.D.; easy turning PTFE stopcocks facilitate turbulence-free entrance of gases, minimizing disturbance of contents
  • Polycarbonate construction is resistant to most common chemicals, as well as thermal stress; wipes clean easily
  • Includes two removable, polypropylene, 36.5cm (14.5 in.) and 22.5cm (9 in.) diameter shelves
  • Side handles provide easy portability and can be used as a place to affix tamper-evident seals
  • Gas ports can be closed to use desiccator with standard desiccant cartridges (08-594-17C, 08-594-17B, or 08-594-14B)
Specifications
Specifications
Transparent
Polycarbonate
11 lb.
Gas-ported Desiccator
45cm
2 gas ports, 2 removable polypropylene shelves (14.5 in. and 9 in.), polypropylene stopcock with PTFE plug and red handle
65L
5kg
18 in.