Learn More
Bel-Art™ SP Scienceware™ Tiered Test Tube Rack, Autoclavable
Double-tiered with solid bottom platform uses minimal space and keeps tubes stable on benchtop
Manufacturer:  Bel-Art™ H188620000
Catalog No. 11-391-304
Add to cart
Description
Description
  • Compact rack keeps test tubes upright and organized
  • Includes convenient holes for mounting to benchtop
  • Made of durable, high density polypropylene; heat and chemical resistant
  • Autoclavable at 121°C (250°F)
Specifications
Specifications
Autoclavable
14 mm
10-14mm tubes
Polypropylene
Tube Rack
White
23.9 x 6.4 x 6.6cm (9.4 x 2.5 x 2.6 in.)
10 to 14 mm Tubes
16