Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Touch Free™ Automatic Waste Cans

Automatically open and close without a touch, protecting users from waste receptacle contact

Manufacturer:  Bel-Art™ F132020022

 View more versions of this product

Catalog No. 03-409-55


Add to cart

Includes: Each unit is supplied with a brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications).

Requires : Requires four “D” batteries (not included) which will provide approximately 4000 openings. All cans should be lined with disposable trash or biohazard bags (sold separately). Batteries and bags available on www.fishersci.com.

Description

Description

Automatically open and close without a touch, protecting users from waste receptacle contact

  • When a hand or solid object is moved over the top, waste can detects motion, opens lid to accept waste, and then closes automatically
  • Fitted well helps to reduce odor and airborne contamination
  • Durable, dependable construction accommodates Scienceware™ liners, biohazard liners or commercially available trash bags
  • ON-OFF switch deactivates AUTO-OPEN/CLOSE function

2 Gallon (Cat. No. 11-394-451)

  • Ideal for benchtop use
  • Removable rigid autoclavable polypropylene liner will not leak or rust
  • Body and lid made from easy-to-clean, molded ABS plastic
  • White pail with white top
  • Line with 14 x 19 in. biohazard disposal bags or suitable size standard trash bags
7.3 Gallon (Cat. No. 11-394-452)
  • Pail made from polypropylene with an ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • Gray pail with gray top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
7.3 Gallon (Cat. No. 03-409-55)
  • Pail made from polypropylene with an ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
  • Red pail with red top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
12 Gallon (Cat. No. 11-394-453)
  • Small footprint enables a fit into tight spaces
  • Stainless-steel pail with black ABS plastic top
  • Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags

Prior to use, cans should be appropriately labeled with one of the included labels to indicate the type of waste to be placed inside the can.

Specifications

Specifications

7.3 gal.
Red
22.5 in.
Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications)
36.8 cm
Automatic Waste Can
26.7 cm
27.6 L
36.8 x 26.7 x 57.1 cm (14.4 x 10.5 x 22.5 in.)
57.1 cm
14.4 in.
ABS
10.5 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit