Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Two-Color Wash Bottles - Safety-Vented™ and Safety Labeled, Wide-Mouth

Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Ethanol
Isopropanol
LYOB (Label Your Own Bottle)
Methanol
Methyl Ethyl Ketone
Use only with solvents compatible with LDPE.
Use only with solvents compatible with LDPE.

A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles

Volatiles are vented eliminating solvent drips. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.

  • Valve eliminates drips; allows dispensing of wash bottle in an upright or inverted position
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Polypropylene closure with leakproof design; color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

Order Info

8 oz. (250mL)

Capacity 16 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Chemical Label Methyl Ethyl Ketone
Closure Material Polypropylene
For Use With (Application) Methyl Ethyl Ketone
Includes Venting Solvent Labled Wash Bottle
Material LDPE
Autoclavable No
Closure Color Green
Closure Size 53mm
Mouth Wide
Show More Show Less