Bel-Art™ SP Scienceware™ Two-Piece Stopcocks

Two-piece design snaps together forming a tight seal, and the free-turning key needs no lubrication

Manufacturer: Bel-Art™ F309170000

 View more versions of this product

Catalog No. 22-088419

  Check Availability
Add to Cart

    Specifications

    Connectors Barbed
    Length (Metric) 98mm
    Fits Tubing ID 12.7mm
    Material Polyethylene
    Plug Type 2-piece
    For Use With Flexible plastic or rubber tubing
    Length (English) 3.85 in.
    Pressure 7psi

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit