Learn More
Bel-Art™ SP Scienceware™ Vikem™ Vinyl Coated Lead Sheets
For stabilizing laboratory equipment in water baths or any application where extra weight is needed
Manufacturer:  Bel-Art™ F183210212
Catalog No. 22-306728
Add to cart
Description
Description
  • May be bent into cylinders, “S” curves, “U” curves or twists
  • Cushioned, non-skid vinyl coating ensures scratching labware equipment
  • 2.2mm (0.085 in.) thick
  • Dimensions: 2 x 12 in. (5.1 x 30.5cm)
Specifications
Specifications
To stabilizing laboratory equipment in water baths or any application where extra weight is needed
Lead
0.22cm
Lead Sheet
0.08 in.