Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles

Click to view available options
Capacity:
16 oz., 500 mL
32 oz., 1000 mL
For Use With (Application):
Acetone
Dichloromethane
Ethanol
Ethyl Acetate
Methyl Ethyl Ketone
Saline
Sodium Hypochlorite

Includes

Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information

Easy-to-read labeling printed bottle promoting safe working environment

  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Made from solvent-resistant low-density polyethylene
  • Polypropylene closure with leakproof design
  • Pictogram instructions promote universal safety regardless of setting

Use LYOB only with solvents compatible with LDPE.

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Closure Material Polypropylene
Color Clear
For Use With (Application) Methyl Ethyl Ketone
Material LDPE
Autoclavable No
Closure Size 53mm
Mouth Wide