Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles Available on GSA/VA Contract for Federal Government customers only.

Easy-to-read labeling printed bottle promoting safe working environment

Manufacturer:  Bel-Art™ F116460627

 View more versions of this product

Catalog No. 14-512-38



Add to cart

Includes: Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information

Description

Description

  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Made from solvent-resistant low-density polyethylene
  • Polypropylene closure with leakproof design
  • Pictogram instructions promote universal safety regardless of setting

Use LYOB only with solvents compatible with LDPE.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Specifications

Specifications

16 oz., 500mL
16 oz.
Clear
LDPE
Blue
Wide
500 mL
Polypropylene
Sodium Hypochlorite
No
43mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit